Learn More
Succinic Anhydride, 99%
Succinic anhydride, 99%, C4H4O3, CAS Number-108-30-5, 2,5-diketotetrahydrofuran, 2,5-furandione, dihydro, butanedioic anhydride, dihydro-2,5-furandione, dihydrofuran-2,5-dione, rikacid sa, succinic acid anhydride, succinic anhydride, succinyl anhydride, succinyl oxide, 1kg, 17,407, 261.0 deg.C | CAS: 108-30-5 | C4H4O3 | 100.07 g/mol
Supplier: Thermo Scientific Chemicals 158760051
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Succinic anhydride | |
| 108-30-5 | |
| 5 kg | |
| 99% | |
| MFCD00005525 | |
| 17,407 | |
| 15,9 | |
| Solubility in water: 62g/L (20°C). Other solubilities: soluble in chloroform,carbon tetrachloride and,alcohol,slightly soluble in ether,20g/L benzene (20°C) | |
| O=C1CCC(=O)O1 | |
| 100.07 | |
| CHEBI:36595 | |
| 99% |
| 116.0°C to 120.0°C | |
| 261.0°C | |
| Plastic drum | |
| C4H4O3 | |
| 3 mPa.s (125°C) | |
| 04,468 | |
| succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa | |
| RINCXYDBBGOEEQ-UHFFFAOYSA-N | |
| oxolane-2,5-dione | |
| 7922 | |
| 100.07 |
Chemical Identifiers
| 108-30-5 | |
| 100.07 | |
| RINCXYDBBGOEEQ-UHFFFAOYSA-N | |
| 7922 | |
| oxolane-2,5-dione |
| C4H4O3 | |
| MFCD00005525 | |
| succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa | |
| CHEBI:36595 | |
| O=C1CCC(=O)O1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately
GHS Signal Word: Danger
EINECSNumber : 203-570-
RUO – Research Use Only