missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Succinic Anhydride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC158762500

 View more versions of this product

Catalog No. AC158762500



Specifications

Specifications

Succinic anhydride
Plastic bottle
261.0°C
250g
99%
MFCD00005525
04,468
2,5-diketotetrahydrofuran
RINCXYDBBGOEEQ-UHFFFAOYSA-N
oxolane-2,5-dione
7922
100.07
147°C
3 mPa.s (125°C)
116.0°C to 120.0°C
108-30-5
C4H4O3
17,407
15,9
Solubility in water: 62g/L (20°C). Other solubilities: soluble in chloroform,carbon tetrachloride and,alcohol,slightly soluble in ether,20g/L benzene (20°C)
C1CC(=O)OC1=O
100.07
CHEBI:36595
99%

Chemical Identifiers

108-30-5
100.07
RINCXYDBBGOEEQ-UHFFFAOYSA-N
7922
oxolane-2,5-dione
C4H4O3
MFCD00005525
2,5-diketotetrahydrofuran
CHEBI:36595
C1CC(=O)OC1=O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately

GHS Signal Word: Danger

EINECSNumber : 203-570-

SDS Documents