missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Succinic Anhydride, 99%, ACROS Organics™

Manufacturer:  Acros Organics 158760050

 View more versions of this product

Catalog No. AC158760050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Succinic anhydride
Glass Bottle
116.0°C to 120.0°C
108-30-5
C4H4O3
succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
C1CC(=O)OC1=O
100.073
CHEBI:36595
99%
147°C
261.0°C
5g
99%
MFCD00005525
RINCXYDBBGOEEQ-UHFFFAOYSA-N
oxolane-2,5-dione
7922
100.07

Chemical Identifiers

108-30-5
100.073
RINCXYDBBGOEEQ-UHFFFAOYSA-N
7922
oxolane-2,5-dione
C4H4O3
MFCD00005525
succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
CHEBI:36595
C1CC(=O)OC1=O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents