missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Succinic acid, disodium salt hexahydrate, 97%, ACROS Organics™

Manufacturer:  Acros Organics 158755000

 View more versions of this product

Catalog No. AC158755000



Specifications

Specifications

Succinic acid, disodium salt hexahydrate
6106-21-4
C4H16Na2O10
02, 606
butanedioic acid disodium salt
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
270.142
CHEBI:63686
Liquid
10ppm max.
0.005% max.
Glass bottle
120.0°C
Brown to Yellow
97%
97%
0.02% max.
15, 8808
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
disodium;butanedioate;hexahydrate
3083938
270.13
97%
Authentic
37.0 % to 41.0 % (90°C)
Solubility in water: 270g/L water (25°C). Other solubilities: insoluble in alcohol
7 to 9 (0.4M soln.)
500g

Chemical Identifiers

6106-21-4
270.142
butanedioic acid disodium salt
CHEBI:63686
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
C4H16Na2O10
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
3083938
disodium;butanedioate;hexahydrate
SDS Documents