Please login to your online account to display your discounted pricing

Succinic Acid, Disodium Salt, 6-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 5kg
Packaging Poly Pail

Manufacturer: avantor performance  V10305

 View more versions of this product

Catalog No. 02004393

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 270.15
Chemical Name or Material Succinic Acid, Disodium Salt
Name Note 6-Hydrate
Molecular Formula NaOCOCH2CH2COONa·6H2O
Quantity 5kg
Purity Grade BAKER ANALYZED™ Reagent
CAS 6106-21-4
Packaging Poly Pail

Specifications

Chemical Name or Material Succinic Acid, Disodium Salt
CAS 6106-21-4
Name Note 6-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOCOCH2CH2COONa·6H2O
Formula Weight 270.15