missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Succinic acid, ACS reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 219550010

 View more versions of this product

Catalog No. AC219550010


 

Specifications Description & Specifications

Specifications

Succinic acid
110-15-6
Crystalline Powder
99+%
C4H6O4
118.09
Butanedioic acid
0.01% max.
0.001% max.
0.001% max.
0.02% max. ()
White
p.a.
ACS Reagent Grade
≥99.0%
0.001% max.
HO2CCH2CH2CO2H
MFCD00002789
5ppm max.
0.0005% max.
Plastic bottle
0.003% max.
185.0° to 191.0°C
1kg

Chemical Identifiers

110-15-6
C4H6O4
118.09
Butanedioic acid
≥99.0%
99+%
HO2CCH2CH2CO2H
MFCD00002789
Succinic acid, p.a.
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-740-4

RTECSNumber : WM4900000

TSCA : TSCA