Please login to your online account to display your discounted pricing

Succinic acid, 99+%, Alfa Aesar™

$198.89 - $198.89

Chemical Identifiers

Chemical Name or Material Succinic acid
CAS 110-15-6
Molecular Formula HO2CCH2CH2CO2H
Formula Weight 118.09
MDL Number MFCD00002789
Beilstein 1754069
Merck Index 14,8869
Synonym Butanedioic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA120840E

  SDS
alfa aesar
A120840E
Each for $198.89
Description & Specifications

Specifications

Melting Point 186° to 188°C
Formula Weight 118.09
Quantity 2500g
Chemical Name or Material Succinic acid
TSCA TSCA
Boiling Point 235°C
Flash Point 206°C (402°F)
Assay ≥99%
Molecular Formula HO2CCH2CH2CO2H
MDL Number MFCD00002789
CAS 110-15-6
Beilstein 1754069
Merck Index 14,8869
Synonym Butanedioic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H335-H315;
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number WM4900000

Applications
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

Solubility
Soluble in ethanol, ethyl ether, acetone and methanol. Insoluble in toluene, benzene, carbon disulfide, carbon tetrachloride and petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315;
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.