missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Succinate, 0.2M buffer soln., pH 5.5

$59.51 - $93.45

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00002789
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61100AK View Documents Alfa Aesar™
J61100AK
250mL Each for $59.51
 
AAJ61100AP View Documents Alfa Aesar™
J61100AP
500mL Each for $93.45
 
Specifications

Chemical Identifiers

150-90-3
162.052
ZDQYSKICYIVCPN-UHFFFAOYSA-L
CHEBI:63675
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
C4H4Na2O4
MFCD00002789
9020
disodium;butanedioate

Specifications

150-90-3
MFCD00002789
ZDQYSKICYIVCPN-UHFFFAOYSA-L
disodium;butanedioate
9020
Liquid
5.5
250mL
C4H4Na2O4
0.2M buffer soln.
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
162.052
CHEBI:63675
Odorless
Colorless
Succinate buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C