Please login to your online account to display your discounted pricing

Alfa Aesar™ Succinamide, 97%

$28.76 - $190.09

Chemical Identifiers

Chemical Name or Material Succinamide
CAS 110-14-5
Molecular Formula H2NC(O)CH2CH2C(O)NH2
Formula Weight 116.12g/mol
MDL Number MFCD00008042
Beilstein 1753983
Merck Index 14,8866
Synonym Butanedioic acid diamide; Succinic diamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1213514

 
alfa aesar™
A12135-14
25g Each for $28.76

AAA1213530

 
alfa aesar™
A12135-30
250g Each for $190.09
Description & Specifications

Specifications

Melting Point ≈263°C (dec.)
Quantity 25g
Chemical Name or Material Succinamide
CAS 110-14-5
Assay 97%
Molecular Formula H2NC(O)CH2CH2C(O)NH2
Formula Weight 116.12g/mol
MDL Number MFCD00008042
Beilstein 1753983
Merck Index 14,8866
Synonym Butanedioic acid diamide; Succinic diamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.