Please login to your online account to display your discounted pricing

Succinaldehyde disodium bisulfite, 92%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  419750050

 View more versions of this product

Catalog No. AC419750050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
Quantity 5g
Chemical Name or Material Succinaldehyde disodium bisulfite
Name Note 92%
CAS 5450-96-4
Physical Form Beige powder
Assay 90% min.
Assay Percent Range 92%
Molecular Formula C4H8Na2O8S2
Linear Formula NaO3SCH(OH)CH2CH2CH(OH)SO3Na
Formula Weight 294.20g/mol
MDL Number MFCD00024982
RTECS Number EK3585000
EINECS Number 226-676-9
TSCA TSCA

Specifications

Chemical Name or Material Succinaldehyde disodium bisulfite
Name Note 92%
CAS 5450-96-4
Assay 90% min.
Assay Percent Range 92%
Molecular Formula C4H8Na2O8S2
Linear Formula NaO3SCH(OH)CH2CH2CH(OH)SO3Na
Formula Weight 294.20g/mol
MDL Number MFCD00024982