Please login to your online account to display your discounted pricing

Succinaldehyde disodium bisulfite, 92%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Succinaldehyde disodium bisulfite
Name Note 92%
CAS 5450-96-4
Assay 90% min.
Assay Percent Range 92%
Molecular Formula C4H8Na2O8S2
Linear Formula NaO3SCH(OH)CH2CH2CH(OH)SO3Na
Formula Weight 294.20g/mol
MDL Number MFCD00024982
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC419750050

  SDS
thermo fisher scientific
419750050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
Quantity 5g
Chemical Name or Material Succinaldehyde disodium bisulfite
Name Note 92%
CAS 5450-96-4
Physical Form Beige powder
Assay 90% min.
Assay Percent Range 92%
Molecular Formula C4H8Na2O8S2
Linear Formula NaO3SCH(OH)CH2CH2CH(OH)SO3Na
Formula Weight 294.20g/mol
MDL Number MFCD00024982
RTECS Number EK3585000
EINECS Number 226-676-9
TSCA TSCA