Please login to your online account to display your discounted pricing

Alfa Aesar™ Substance P, Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67854ODL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67854ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Regulatory Status RUO
Quantity 3 vials, 500 pmol/vial
Purification Method HPLC
Sequence RPKPQQFFGLM
Product Type Substance P, Certified Mass Spec Grade
Molecular Weight 1348.60
Format Lyophilized powder