missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Suberic acid bis(N-hydroxysuccinimide ester), 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC426020010

 View more versions of this product

Catalog No. AC426020010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Suberic acid bis(N-hydroxysuccinimide ester)
1g
96% min. (HPLC)
MFCD00049059
ZWIBGKZDAWNIFC-UHFFFAOYSA-N
bis(2,5-dioxopyrrolidin-1-yl) octanedioate
100658
97%
Glass bottle
68528-80-3
C16H20N2O8
2,5-pyrrolidinedione, 1,1'-1,8-dioxo-1,8-octanediyl bis oxy bis, bicl100, bis 2,5-dioxopyrrolidin-1-yl octanedioate, di n-succinimidyl suberate, disuccinimidyl suberate, disuccinimidylsuberate, dsis, n-hydroxysuccinimide suberic acid ester, nhs-sa, suberic acid bis n-hydroxysuccinimide ester
C1CC(=O)N(C1=O)OC(=O)CCCCCCC(=O)ON2C(=O)CCC2=O
368.34
368.34

Chemical Identifiers

68528-80-3
368.34
ZWIBGKZDAWNIFC-UHFFFAOYSA-N
100658
C1CC(=O)N(C1=O)OC(=O)CCCCCCC(=O)ON2C(=O)CCC2=O
C16H20N2O8
MFCD00049059
2,5-pyrrolidinedione, 1,1'-1,8-dioxo-1,8-octanediyl bis oxy bis, bicl100, bis 2,5-dioxopyrrolidin-1-yl octanedioate, di n-succinimidyl suberate, disuccinimidyl suberate, disuccinimidylsuberate, dsis, n-hydroxysuccinimide suberic acid ester, nhs-sa, suberic acid bis n-hydroxysuccinimide ester
bis(2,5-dioxopyrrolidin-1-yl) octanedioate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents