Please login to your online account to display your discounted pricing

SU 9516, Tocris

Potent cdk2 inhibitor

$181.19 - $745.63

Chemical Identifiers

Chemical Name or Material SU 9516
Assay Percent Range >99%
Molecular Formula C13H11N3O2
Formula Weight 241.25
Synonym (Z)-1, 3-Dihydro-3-(1H-imidazol-4-ylmethylene)-5-methoxy-2H-indol-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

290710

 
r & d systems
RDS290710
10mg Each for $181.19

290750

 
r & d systems
RDS290750
50mg Each for $745.63
Description & Specifications

Specifications

Formula Weight 241.25
Quantity 10mg
Chemical Name or Material SU 9516
Assay Percent Range >99%
Molecular Formula C13H11N3O2
CAS 377090-84-1
Synonym (Z)-1, 3-Dihydro-3-(1H-imidazol-4-ylmethylene)-5-methoxy-2H-indol-2-one
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C