missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-Styrene oxide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0782122

 View more versions of this product

Catalog No. AAL0782122



Description

Description

Styrene oxide is used as an intermediate in medicine and perfume.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Styrene oxide is used as an intermediate in medicine and perfume.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

(+/-)-Styrene oxide
-37°C
193°C to 196°C
C8H8O
MFCD00005121
UN2810
styrene oxide, phenyloxirane, 1,2-epoxyethylbenzene, phenylethylene oxide, epoxystyrene, styrene epoxide, styryl oxide, oxirane, phenyl, 1-phenyloxirane, phenethylene oxide
AWMVMTVKBNGEAK-UHFFFAOYSA-N
2-phenyloxirane
7276
120.15
96-09-3
1.053
74°C (165°F)
1.535
100 g
108582
Soluble in water.
C1C(O1)C2=CC=CC=C2
120.151
CHEBI:17907
≥98%

Chemical Identifiers

96-09-3
120.151
AWMVMTVKBNGEAK-UHFFFAOYSA-N
7276
2-phenyloxirane
C8H8O
MFCD00005121
styrene oxide, phenyloxirane, 1,2-epoxyethylbenzene, phenylethylene oxide, epoxystyrene, styrene epoxide, styryl oxide, oxirane, phenyl, 1-phenyloxirane, phenethylene oxide
CHEBI:17907
C1C(O1)C2=CC=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302-H312-H319-H227
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Combustible liquid.

P201-P202-P210-P235-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P333+P313-P362-P370+P378q-P501c

H227-H312-H315-H317-H319-H331-H340-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 202-476-7

RTECSNumber : CZ9625000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only