missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Styrene/maleic anhydride copolymer, containing monomethyl ester, ACROS Organics™

Chemical Identifiers

CAS 31959-78-1
MDL Number MFCD00134011
Chemical Name or Material Styrene/maleic anhydride copolymer
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC190912500 SDS Thermo Fisher Scientific
190912500
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

31959-78-1
MFCD00134011
Styrene/maleic anhydride copolymer

Specifications

31959-78-1
MFCD00134011
2% max.
250g
Crystalline Powder
Authentic
White to light yellow
Styrene/maleic anhydride copolymer

Safety and Handling

RTECSNumber : ON4240000

TSCA : TSCA