Please login to your online account to display your discounted pricing

Styrene/maleic anhydride copolymer, containing monomethyl ester, ACROS Organics™


Quantity 250g

Manufacturer: thermo fisher scientific  190912500

 View more versions of this product

Catalog No. AC190912500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Quantity 250g
Color White to light yellow
RTECS Number ON4240000
TSCA TSCA
Loss on Drying 2% max.
Infrared Spectrum Authentic
Chemical Name or Material Styrene/maleic anhydride copolymer
CAS 31959-78-1
MDL Number MFCD00134011

Specifications

Chemical Name or Material Styrene/maleic anhydride copolymer
CAS 31959-78-1
MDL Number MFCD00134011