Please login to your online account to display your discounted pricing

Styrene-d8, for NMR, 98+% atom D, stabilized, ACROS Organics™

$467.04 - $467.04

Chemical Identifiers

Chemical Name or Material Styrene-d8
Name Note 98+ atom% D
CAS 19361-62-7
Purity Grade For NMR
Molecular Formula C8D8
Linear Formula C6D5CD=CD2
Formula Weight 112.22
MDL Number MFCD00044231
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC175390050

  SDS
thermo fisher scientific
175390050
Glass bottle 5g Each for $467.04
Description & Specifications

Specifications

Chemical Name or Material Styrene-d8
Name Note 98+ atom% D
CAS 19361-62-7
Deuterium (D) 98.0% min.
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
EINECS Number 242-995-6
Stabilizer 0.5% 4-tert.-butylcatechol
TSCA TSCA
Water 0.05% max. (Coulometric)
Density 0.9700g/mL
Purity Grade For NMR
Physical Form Liquid
Quantity 5g
Color Clear colorless
Molecular Formula C8D8
Linear Formula C6D5CD=CD2
Formula Weight 112.22
MDL Number MFCD00044231

For NMR

Safety and Handling

Danger

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.