Please login to your online account to display your discounted pricing

Styrene-D8, deuteration degree min. 98% for NMR spectroscopy, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 1mL
Packaging Glass ampoule

Manufacturer: emd millipore corporation  8150610001

 View more versions of this product

Catalog No. M8150610001

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Styrene-D8
CAS 19361-62-7
Odor Sweet
Packaging Glass ampoule
Physical Form Liquid
Quantity 1mL
Assay Percent Range 98%
Color Colorless
Molecular Formula C8D8 (Hill)
Formula Weight 112.2
Synonym Phenylethylene

Specifications

Chemical Name or Material Styrene-D8
CAS 19361-62-7
Physical Form Liquid
Assay Percent Range 98%
Molecular Formula C8D8 (Hill)
Formula Weight 112.2
Synonym Phenylethylene