missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Styrene-D8, deuteration degree min. 98% for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$256.85 - $256.85

Chemical Identifiers

CAS 19361-62-7
Physical Form Liquid
Assay Percent Range 98%
Molecular Formula C8D8 (Hill)
Formula Weight 112.2
Synonym Phenylethylene
Chemical Name or Material Styrene-D8
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M8150610001 Emd Millipore Corporation
8150610001
Glass ampoule 1mL Each for $256.85
Specifications Description & Specifications

Chemical Identifiers

19361-62-7
98%
112.2
Styrene-D8
Liquid
C8D8 (Hill)
Phenylethylene

Specifications

19361-62-7
Glass ampoule
1mL
Colorless
112.2
Styrene-D8
Sweet
Liquid
98%
C8D8 (Hill)
Phenylethylene