Please login to your online account to display your discounted pricing

Styrene-D8, deuteration degree min. 98% for NMR spectroscopy, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Packaging Glass ampoule

Manufacturer: emd millipore corporation  8.15061.0010

 View more versions of this product

Catalog No. M8150610010

  • / Each
 

Description & Specifications

Chemical Identifiers

Odor Sweet
Packaging Glass ampoule
Quantity 10mL
Color Colorless
Chemical Name or Material Styrene-D8
CAS 19361-62-7
Physical Form Liquid
Assay Percent Range 98%
Molecular Formula C8D8 (Hill)
Formula Weight 112.2g/mol
Synonym Phenylethylene

Specifications

Chemical Name or Material Styrene-D8
CAS 19361-62-7
Physical Form Liquid
Assay Percent Range 98%
Molecular Formula C8D8 (Hill)
Formula Weight 112.2g/mol
Synonym Phenylethylene