Please login to your online account to display your discounted pricing

Alfa Aesar™ Styrene-d8, 98% (Isotopic) stab. with 4-tert-butylcatechol

Chemical Identifiers

Chemical Name or Material Styrene-d8
CAS 19361-62-7
Name Note Stabilized with 4-tert-butylcatechol, Isotopic
Molecular Formula C8D8
Linear Formula C6D5CD=CD2
Formula Weight 112.2
MDL Number MFCD00044231
UN Number UN2055
Beilstein 2327045
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4366706

 
alfa aesar™
4366706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.2
Packaging Ampule
Chemical Name or Material Styrene-d8
TSCA TSCA
Sensitivity Air Sensitive
Density 0.979g/mL
Name Note Stabilized with 4-tert-butylcatechol, Isotopic
Boiling Point 50° to 51°C (25mmHg)
Physical Form Liquid
Assay 98%
Quantity 5g
Color Colorless
Molecular Formula C8D8
Flash Point 31°C (88°F)
Linear Formula C6D5CD=CD2
Refractive Index 1.5445
MDL Number MFCD00044231
CAS 19361-62-7
UN Number UN2055
Beilstein 2327045
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H332-H315-H319
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Recommended Storage 2°C to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H332-H315-H319
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

2°C to 8°C

  • Ampouled under argon