missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Styrene, 99.5% stab. with 4-tert-butylcatechol

Manufacturer:  Alfa Aesar™ 16199AD

 View more versions of this product

Catalog No. AA16199AD



Specifications

Specifications

Styrene
31° (88°F)
1.5460
145°C to 146°C
-31°C
neutral
100-42-5
MFCD00008612
1071236
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
C=CC1=CC=CC=C1
104.152
CHEBI:27452
Liquid
Technical
0.906g/mL
Pungent
Light Sensitive
Colorless
50mL
Stabilized with 4-tert-butylcatechol
C8H8
UN2055
14,8860
PPBRXRYQALVLMV-UHFFFAOYSA-N
styrene
7501
104.15
99.5%

Chemical Identifiers

100-42-5
104.152
PPBRXRYQALVLMV-UHFFFAOYSA-N
7501
styrene
C8H8
MFCD00008612
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
CHEBI:27452
C=CC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H372-H304-H361-H226-H332-H315-H319-H335
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED

RTECSNumber : WL3675000

TSCA : TSCA

storageNote1 : 2°C to 8°C

SDS Documents