Please login to your online account to display your discounted pricing

Styrene, 99.5%, for analysis, stabilized, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  220532500

 View more versions of this product

Catalog No. AC220532500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Physical Form Liquid
Formula Weight 104.15
Quantity 250mL
Color Clear
TSCA TSCA
Purity Grade For analysis
EINECS Number 202-851-5
RTECS Number WL3675000
Density 0.9060g/mL
Beilstein 05, 474
Merck Index 15, 8990
Peroxides 0.01% max. (as H2O2)
Polymer 0.001% max.
CAS 98-29-3
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Stabilizer 10 to 20ppm p-tert-butylcatechol
Refractive Index 1.5460 to 1.5480 (20°C, 589nm)
Aldehyde 0.02% max.
Infrared Spectrum Authentic
Synonym Ethenylbenzene
Linear Formula C6H5CH=CH2
MDL Number MFCD00008612
CAS 100-42-5
Molecular Formula C8H8
Chemical Name or Material Styrene
Name Note p.a.
Assay Percent Range 99.5%

Specifications

Chemical Name or Material Styrene
Name Note p.a.
CAS 100-42-5
CAS 98-29-3
Purity Grade For analysis
Assay ≥99.5%
Assay Percent Range 99.5%
Molecular Formula C8H8
Linear Formula C6H5CH=CH2
Formula Weight 104.15
MDL Number MFCD00008612
Synonym Ethenylbenzene
Accessories

Danger

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.

SDS & Safety Documents