Student Optical Bench Accessories
For a variety of physics applications using lenses and mirrors.
$3.80 - $3.80
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S42307 Fisher Science Education
EMI6008
Each for $3.80
Specifications
Description
Lens or Mirror Support
  • Durable, nickel-plated steel construction for long life
  • Metal lens/mirror supports with pointed marker located directly below the slot
Incandescent Lamp
  • Miniature, screw-type base for low-voltage lamps
  • Mounted on a support with a marker directly below the geometric center of the lamp
  • Suitable for use with miniature lamps (1.5V, 0.15A; or 6V, 0.5A)
  • Porcelain base has a nickel-plated mounting with a pointer
  • Geometric center of the lamp is 3 in. (7.5cm) above the meter rule