Please login to your online account to display your discounted pricing

Student-Grade Filter Paper


Perfect for routine lab use.

Manufacturer: fisher science education  FP3

 View more versions of this product

Catalog No. S60173

  • / Pack of 1250 Each
 

Description & Specifications

Specifications

Grade Student Grade
Diameter (Metric) 12.5cm
Flow Rate Medium
Quantity 1250 per pack
Format Circle