Please login to your online account to display your discounted pricing

Student-Grade Filter Paper

Perfect for routine lab use.

$182.25 - $182.25

Specifications

Format Circle
Flow Rate Medium
Grade Student Grade
Quantity 1250 per pack
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Price Quantity    

S60173

 
fisher science education
FP3
12.5cm Pack of 1250 Each for $182.25
Description & Specifications

Specifications

Format Circle
Flow Rate Medium
Grade Student Grade
Quantity 1250 per pack

  • Supplied in handy dispensing box
  • Contains 12 boxes of 100 circles and 1 polyethylene bag of 50 circles (total of 1250 pieces)
  • Medium retention and flowrate

Note:Paper fits our largest porcelain Büchner Funnel (S34597-1)