Please login to your online account to display your discounted pricing
Student Cell Kits
Build a voltaic, storage, coulometer, dry or Daniel cell.
$36.40 - $40.35
Specifications
Product Type Chemistry Kit
View More Specs
Includes: 
Comprehensive Kit
  • 8 oz. (0.23kg) opaque polypropylene cell; porous cup; instructions; and labeled electrodes: Copper (2), Zinc (2), Aluminum (1), Carbon (1), Iron (1) and Nickel (1)
Basic Kit
  • Only parts and instructions to build the voltaic cell
Requires:  Acids
Products
Catalog Number Mfr. No. Includes Price Quantity    
S94059
fisher science education
10-180
Comprehensive: 8 oz. (0.23kg) opaque polypropylene cell, porous cup, instructions; and labeled electrodes, 2 copper, 2 zinc, aluminum, carbon, iron and nickel Each for $36.40
S94060
fisher science education
10-175
Basic: Only parts and instructions to build the voltaic cell Each for $40.35
Description & Specifications
Specifications
Product Type Chemistry Kit
  • Screw-on plastic rim has two brass electrode holders that adjust to hold round or flat electrodes
  • Cell body has a low profile and wide base to resist spills