missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Strychnine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158951000

 View more versions of this product

Catalog No. AC158951000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(-)-Strychnine
57-24-9
100.0
White
98%
C21H22N2O2
100 g
strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos
O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61
334.42
CHEBI:28973
98%
98%
97.5
284.0°C to 286.0°C
Authentic
Glass bottle
MFCD00005941
15, 8985
QMGVPVSNSZLJIA-FVWCLLPLSA-N
(4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
441071
334.41
Fine Crystalline Powder

Chemical Identifiers

57-24-9
334.42
QMGVPVSNSZLJIA-FVWCLLPLSA-N
441071
(4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
C21H22N2O2
MFCD00005941
strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos
CHEBI:28973
O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-319-7

RTECSNumber : WL2275000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only