missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium zirconium oxide, 99.3% (metals basis)

Manufacturer:  Alfa Aesar™ 1234630

 View more versions of this product

Catalog No. AA1234630



Specifications

Specifications

Strontium zirconium oxide
(metals basis)
MFCD00049555
FCCTVDGKMTZSPU-UHFFFAOYSA-N
strontium;dioxido(oxo)zirconium
82852
Powder
5.5g/mL
Odorless
White
250g
12036-39-4
O3SrZr
strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate
[O-][Zr](=O)[O-].[Sr+2]
226.841
226.84
99.3%
−325 Mesh
9000°C
2700°C

Chemical Identifiers

12036-39-4
226.841
FCCTVDGKMTZSPU-UHFFFAOYSA-N
82852
[O-][Zr](=O)[O-].[Sr+2]
O3SrZr
MFCD00049555
strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate
strontium;dioxido(oxo)zirconium
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents