missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Strontium zirconium oxide, 99.3% (metals basis), Thermo Scientific Chemicals
$80.85 - $281.23
Chemical Identifiers
CAS | 12036-39-4 |
---|---|
Molecular Formula | O3SrZr |
Molecular Weight (g/mol) | 226.84 |
MDL Number | MFCD00049555 |
InChI Key | FCCTVDGKMTZSPU-UHFFFAOYSA-N |
Synonym | strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate |
PubChem CID | 82852 |
IUPAC Name | strontium;dioxido(oxo)zirconium |
SMILES | [Sr++].[O-][Zr]([O-])=O |
Description
Mostly as intermediates( primary and secondary ). For chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12036-39-4 | |
226.84 | |
FCCTVDGKMTZSPU-UHFFFAOYSA-N | |
82852 | |
[Sr++].[O-][Zr]([O-])=O |
O3SrZr | |
MFCD00049555 | |
strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate | |
strontium;dioxido(oxo)zirconium |
Specifications
9000°C | |
White | |
Powder | |
O3SrZr | |
strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate | |
[Sr++].[O-][Zr]([O-])=O | |
226.84 | |
226.84 | |
Odorless | |
Strontium zirconium oxide |
2700°C | |
12036-39-4 | |
250 g | |
MFCD00049555 | |
FCCTVDGKMTZSPU-UHFFFAOYSA-N | |
strontium;dioxido(oxo)zirconium | |
82852 | |
99.3% | |
5.5 g/mL | |
(metals basis) |
Safety and Handling
EINECSNumber : 234-847-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only