Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium Titanium Oxide Substrate, 6.25x6.25x0.5mm, Polished One Side, 100 Orientation, Alfa Aesar™

$88.88 - $88.88

Chemical Identifiers

Chemical Name or Material Strontium Titanium Oxide Substrate
CAS 12060-59-2
Name Note Polished One Side
Molecular Formula SrTiO3
MDL Number MFCD00049554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42618KT

 
alfa aesar™
42618-KT
1 ea. Each for $88.88
Description & Specifications

Specifications

Odor Odorless
Quantity 1 ea.
Chemical Name or Material Strontium Titanium Oxide Substrate
Name Note Polished One Side
CAS 12060-59-2
Physical Form White to off-white substrate
Molecular Formula SrTiO3
MDL Number MFCD00049554