missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium Titanium Oxide Substrate, 6.25 x 6.25 x 0.5mm, Polished One Side, 110 Orientation, Alfa Aesar™

100 Orientation

$200.46 - $735.99

Chemical Identifiers

CAS 12060-59-2
Physical Form Substrate
Molecular Formula SrTiO3
MDL Number MFCD00049554
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished One Side
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42617KT SDS Alfa Aesar
42617KT
1 ea. Each for $200.46
AA42617KU SDS Alfa Aesar
42617KU
5 ea. Each for $735.99
Specifications Description & Specifications

Chemical Identifiers

12060-59-2
SrTiO3
Strontium Titanium Oxide Substrate, Polished One Side
Substrate
MFCD00049554

Specifications

12060-59-2
SrTiO3
Odorless
1 ea.
Substrate
MFCD00049554
White to off-white
Strontium Titanium Oxide Substrate, Polished One Side