missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Strontium Titanium Oxide Substrate, 5 x 5 x 1mm, Polished Two Sides, 100 Orientation, Thermo Scientific™
100 Orientation
Supplier: Thermo Scientific Chemicals 42670KT
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Strontium Titanium Oxide Substrate | |
White | |
Substrate | |
O3SrTi | |
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder | |
[Sr++].[O-][Ti]([O-])=O | |
183.48 | |
Odorless |
Polished Two Sides | |
12060-59-2 | |
1 Ea. | |
MFCD00049554 | |
VEALVRVVWBQVSL-UHFFFAOYSA-N | |
strontium;dioxido(oxo)titanium | |
82899 |
Chemical Identifiers
12060-59-2 | |
183.48 | |
VEALVRVVWBQVSL-UHFFFAOYSA-N | |
82899 |
O3SrTi | |
MFCD00049554 | |
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder | |
[Sr++].[O-][Ti]([O-])=O |
Safety and Handling
EINECSNumber : 235-044-1
TSCA : Yes
Recommended Storage : Ambient temperatures