Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium Titanium Oxide Substrate, 5 x 5 x 0.5mm, Polished Two Sides, 111 Orientation, Alfa Aesar™

111 Orientation

$153.69 - $562.03

Chemical Identifiers

Chemical Name or Material Strontium Titanium Oxide Substrate
Name Note Polished Two Sides
CAS 12060-59-2
Physical Form Substrate
Molecular Formula SrTiO3
MDL Number MFCD00049554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42594KT

  SDS
alfa aesar™
42594KT
1 ea. Each for $153.69

AA42594KU

  SDS
alfa aesar™
42594KU
5 ea. Each for $562.03
Description & Specifications

Specifications

Physical Form Substrate
Quantity 1 ea.
Color White to off-white
Chemical Name or Material Strontium Titanium Oxide Substrate
CAS 12060-59-2
Molecular Formula SrTiO3
MDL Number MFCD00049554
Odor Odorless
Name Note Polished Two Sides