Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium Titanium Oxide Substrate, 5 x 5 x 0.5mm, Polished Two Sides, 110 Orientation, Alfa Aesar™

$213.96 - $763.93

Chemical Identifiers

Chemical Name or Material Strontium Titanium Oxide Substrate
CAS 12060-59-2
Name Note Polished Two Sides
Molecular Formula SrTiO3
MDL Number MFCD00049554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42669KU

  SDS
alfa aesar™
42669KU
5 ea. Each for $763.93

AA42669KT

  SDS
alfa aesar™
42669KT
1 ea. Each for $213.96
Description & Specifications

Specifications

Physical Form Substrate
Color White to off-white
Quantity 5 ea.
Molecular Formula SrTiO3
CAS 12060-59-2
Chemical Name or Material Strontium Titanium Oxide Substrate
MDL Number MFCD00049554
Name Note Polished Two Sides
Odor Odorless