Please login to your online account to display your discounted pricing

Strontium Titanium Oxide Substrate, 5 x 5 x 0.5mm, Polished Two Sides, 100 Orientation, Alfa Aesar™

$149.93 - $540.28

Chemical Identifiers

Chemical Name or Material Strontium Titanium Oxide Substrate
CAS 12060-59-2
Name Note Polished Two Sides
Molecular Formula SrTiO3
MDL Number MFCD00049554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42668KT

  SDS
alfa aesar™
42668KT
1 ea. Each for $149.93

AA42668KU

  SDS
alfa aesar™
42668KU
5 ea. Each for $540.28
Description & Specifications

Specifications

Physical Form Substrate
Color White to off-white
Quantity 1 ea.
MDL Number MFCD00049554
Chemical Name or Material Strontium Titanium Oxide Substrate
Odor Odorless
Name Note Polished Two Sides
Molecular Formula SrTiO3
CAS 12060-59-2