missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium Titanium Oxide Substrate, 5 x 5 x 0.5mm, Polished One Side, 100 Orientation, Alfa Aesar™

$186.26 - $631.11

Chemical Identifiers

CAS 12060-59-2
Molecular Formula SrTiO3
MDL Number MFCD00049554
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished One Side
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42620KT SDS Alfa Aesar™
42620KT
1 ea. Each for $186.26
AA42620KU SDS Alfa Aesar™
42620KU
5 ea. Each for $631.11
Specifications Description & Specifications

Chemical Identifiers

12060-59-2
SrTiO3
MFCD00049554
Strontium Titanium Oxide Substrate, Polished One Side

Specifications

12060-59-2
SrTiO3
Odorless
1 ea.
Substrate
MFCD00049554
White to off-white
Strontium Titanium Oxide Substrate, Polished One Side