missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium Titanium Oxide Substrate, 10 x 10 x 1mm, Polished Two Sides, 100 Orientation, Alfa Aesar™

100 Orientation

$326.11 - $1,412.15

Chemical Identifiers

CAS 12060-59-2
Physical Form Substrate
Molecular Formula SrTiO3
MDL Number MFCD00049554
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished Two Sides
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA41440KT SDS Alfa Aesar
41440KT
1 ea. Each for $326.11
AA41440KU SDS Alfa Aesar
41440KU
5 ea. Each for $1,412.15
Specifications Description & Specifications

Chemical Identifiers

12060-59-2
SrTiO3
Strontium Titanium Oxide Substrate, Polished Two Sides
Substrate
MFCD00049554

Specifications

12060-59-2
SrTiO3
Odorless
1 ea.
Substrate
MFCD00049554
White to off-white
Strontium Titanium Oxide Substrate, Polished Two Sides