Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium Titanium Oxide Substrate, 10 x 10 x 1mm, Polished One Side, 111 Orientation, Alfa Aesar™

$244.09 - $991.45

Chemical Identifiers

Chemical Name or Material Strontium Titanium Oxide Substrate
CAS 12060-59-2
Name Note Polished One Side
Molecular Formula SrTiO3
MDL Number MFCD00049554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40279KT

  SDS
alfa aesar™
40279KT
1 ea. Each for $244.09

AA40279KU

  SDS
alfa aesar™
40279KU
5 ea. Each for $991.45
Description & Specifications

Specifications

Physical Form Substrate
Color White to off-white
Quantity 1 ea.
Molecular Formula SrTiO3
MDL Number MFCD00049554
Chemical Name or Material Strontium Titanium Oxide Substrate
CAS 12060-59-2
Name Note Polished One Side
Odor Odorless