Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium Titanium Oxide Substrate, 10 x 10 x 1mm, Polished One Side, 110 Orientation, Alfa Aesar™

$327.86 - $1,490.03

Chemical Identifiers

Chemical Name or Material Strontium Titanium Oxide Substrate
CAS 12060-59-2
Name Note Polished One Side
Molecular Formula SrTiO3
MDL Number MFCD00049554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40278KT

 
alfa aesar™
40278KT
1 ea. Each for $327.86

AA40278KU

 
alfa aesar™
40278KU
5 ea. Each for $1,490.03
Description & Specifications

Specifications

Physical Form Substrate
Color White to off-white
Quantity 1 ea.
Chemical Name or Material Strontium Titanium Oxide Substrate
Odor Odorless
Name Note Polished One Side
Molecular Formula SrTiO3
MDL Number MFCD00049554
CAS 12060-59-2