Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium sulfide, 99.995% (metals basis)

Chemical Identifiers

Chemical Name or Material Strontium sulfide
CAS 1314-96-1
Formula Weight 119.68g/mol
MDL Number MFCD00049560
Merck Index 14,8849
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1447306

 
alfa aesar™
14473-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1447314

 
alfa aesar™
14473-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 3.70g/mL
Odor Like rotten eggs
Packaging Ampule
Sensitivity Hygroscopic
Color Gray
Melting Point >2000°C
Quantity 5g
Chemical Name or Material Strontium sulfide
CAS 1314-96-1
Physical Form Powder
Assay 99.995%
Assay Percent Notes (metals basis)
Formula Weight 119.68g/mol
MDL Number MFCD00049560
Merck Index 14,8849
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
Storage Note 1
  • Ampouled under argon
TSCA TSCA

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon