missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium sulfide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 8989509

 View more versions of this product

Catalog No. AA8989509



Specifications

Specifications

Strontium sulfide
(metals basis)
MFCD00049560
strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12, strontium sulfide
XXCMBPUMZXRBTN-UHFFFAOYSA-N
sulfanylidenestrontium
14820
Powder
3.70g/mL
Hygroscopic
>2000°C
1314-96-1
SSr
14,8849
Slightly soluble in water; Decomposes in acids
S=[Sr]
119.68
119.68
99.9%
Like rotten eggs
Gray
10g

Chemical Identifiers

1314-96-1
119.68
XXCMBPUMZXRBTN-UHFFFAOYSA-N
14820
S=[Sr]
SSr
MFCD00049560
strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12, strontium sulfide
sulfanylidenestrontium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents