Learn More
Strontium sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 08989518
Description
Strontium sulfide is used in chemical research and as organic intermediates. It is also used as depilatory. It is also used as Isotope labelled enantiomer of Voriconazole (V760000), an antifungal agent and an ergosterol biosynthesis inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Strontium sulfide | |
1314-96-1 | |
Powder | |
(metals basis) | |
MFCD00049560 | |
strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12 | |
ZEGFMFQPWDMMEP-UHFFFAOYSA-N | |
strontium(2+) sulfanediide | |
14820 | |
99.9% | |
Hygroscopic |
>2000°C | |
Gray | |
50 g | |
SSr | |
14,8849 | |
Slightly soluble in water; Decomposes in acids | |
[S--].[Sr++] | |
119.68 | |
119.68 | |
Like rotten eggs | |
3.70 g/mL |
Chemical Identifiers
1314-96-1 | |
119.68 | |
ZEGFMFQPWDMMEP-UHFFFAOYSA-N | |
14820 | |
[S--].[Sr++] |
SSr | |
MFCD00049560 | |
strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12 | |
strontium(2+) sulfanediide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H301-H314
EINECSNumber : 215-249-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only