missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Strontium sulfate, Puratronic™, 99.9965% (metals basis)

Manufacturer:  Alfa Aesar™ 1287006

 View more versions of this product

Catalog No. AA1287006



Specifications

Specifications

Strontium sulfate
(metals basis)
SrSO4
14,8848
UBXAKNTVXQMEAG-UHFFFAOYSA-L
strontium;sulfate
3084026
Powder
Puratronic™
Odorless
1600°C
7759-02-6
O4SSr
MFCD00036274
strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g, strontium sulfate
[O-]S(=O)(=O)[O-].[Sr+2]
183.68
183.68
99.9965%
3.960g/mL
White
5g

Chemical Identifiers

7759-02-6
183.68
UBXAKNTVXQMEAG-UHFFFAOYSA-L
3084026
[O-]S(=O)(=O)[O-].[Sr+2]
O4SSr
MFCD00036274
strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g, strontium sulfate
strontium;sulfate
Safety and Handling

Safety and Handling

RTECSNumber : WT1210000

TSCA : TSCA

Documents