Please login to your online account to display your discounted pricing

Strontium Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep


Quantity 250mL
Packaging Poly Bottle

Manufacturer: spex certiprep llc  PLSR22T

 View more versions of this product

Catalog No. PLSR22T

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 250mL
DOT Information Nonhazardous
Recommended Storage Ambient
Health Hazard 1 Toxic in contact with skin. Causes serious eye irritation
Purity Grade Assurance Grade
Packaging Poly Bottle
CAS 7440-24-6
CAS 7732-18-5
Chemical Name or Material Strontium Standard
CAS 7697-37-2

Specifications

Chemical Name or Material Strontium Standard
CAS 7440-24-6
CAS 7732-18-5
CAS 7697-37-2
Purity Grade Assurance Grade
Accessories

Toxic in contact with skin. Causes serious eye irritation

Ambient

SDS & Safety Documents