Please login to your online account to display your discounted pricing

Strontium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 125mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  PLSR22Y

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. PLSR2-2Y

  • / Each
 

Description & Specifications

Chemical Identifiers

DOT Information Hazardous
Health Hazard 1 Causes severe skin burns and eye damage
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Recommended Storage Ambient
Quantity 125mL
Solvent or Matrix 2% HNO3
Packaging Triple-leached LDPE bottle
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Purity Grade Assurance Grade
CAS 7732-18-5
CAS 7697-37-2
Chemical Name or Material Strontium Standard
CAS 7440-24-6

Specifications

Chemical Name or Material Strontium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sr1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes severe skin burns and eye damage

Ambient

SDS & Safety Documents