Please login to your online account to display your discounted pricing

Strontium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Sr) Solute: Strontium Carbonate Solvent: Dilute Hydrochloric Acid

Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SS459500

 View more versions of this product

Catalog No. SS459-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg
Viscosity [NULL]
Density 1.0g/cm³
pH 1
Name Note 1000ppm ±1%
MDL Number MFCD00134060
CAS 1633-05-2
CAS 7732-18-5
Concentration 990 to 1010ppm
CAS 7647-01-0
Packaging Poly Bottle
Traceability to NIST 3153a STRONTIUM STANDARD
Chemical Name or Material Strontium, Reference Standard Solution
ChemAlert Storage Symbol White

Specifications

Chemical Name or Material Strontium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 1633-05-2
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134060
Accessories
 

SDS & Safety Documents