Please login to your online account to display your discounted pricing

Strontium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Sr) Solute: Strontium Carbonate Solvent: Dilute Hydrochloric Acid

Quantity 100mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSS459100

 View more versions of this product

Catalog No. SS459-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 0°C
Boiling Point 100°C
Purity Grade Certified Grade
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
pH 1
Density 1.0g/cm³
Color Colorless
Vapor Pressure 14mmHg
Chemical Name or Material Strontium, Reference Standard Solution
MDL Number MFCD00134060
ChemAlert Storage Symbol White
CAS 7647-01-0
Traceability to NIST 3153a STRONTIUM STANDARD
Concentration 990 to 1010ppm
Packaging Poly Bottle
CAS 7732-18-5
Name Note 1000ppm ±1%
CAS 1633-05-2

Specifications

Chemical Name or Material Strontium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 1633-05-2
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134060
Accessories
 

SDS & Safety Documents