Please login to your online account to display your discounted pricing

Strontium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Sr) Solute: Strontium Carbonate Solvent: Dilute Hydrochloric Acid

$113.15 - $287.62

Chemical Identifiers

Chemical Name or Material Strontium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 1633-05-2
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134060
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SS459-500

  SDS
fisher scientific company
SS459500
Poly Bottle 500mL Each for $287.62
Case of 6 Each for $1,725.72

SS459-100

  SDS
fisher scientific company
FLSS459100
Poly Bottle 100mL Each for $113.15
Case of 6 Each for $678.90
Description & Specifications

Specifications

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg
Density 1.0g/cm³
pH 1
Name Note 1000ppm ±1%
MDL Number MFCD00134060
CAS 1633-05-2
CAS 7732-18-5
Concentration 990 to 1010ppm
CAS 7647-01-0
Packaging Poly Bottle
Traceability to NIST 3153a STRONTIUM STANDARD
Chemical Name or Material Strontium, Reference Standard Solution
ChemAlert Storage Symbol White
Safety and Handling